Thermo Scientific Chemicals

3,5-Difluoro-4-methoxybenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: H26526.03
1 g, Each
Thermo Scientific Chemicals

3,5-Difluoro-4-methoxybenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: H26526.03
1 g, Each
Quantity
Catalog number: H26526.03
also known as H26526-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
706786-42-7
IUPAC Name
5-(bromomethyl)-1,3-difluoro-2-methoxybenzene
Molecular Formula
C8H7BrF2O
InChI Key
IQTBMXJVMUSGSL-UHFFFAOYSA-N
SMILES
COC1=C(F)C=C(CBr)C=C1F
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)
≥96.0%
Melting Point (clear melt)
36.5-42.5?C

Description

3,5-Difluoro-4-methoxybenzyl bromide is useful for research purposes. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluoro-4-methoxybenzyl bromide is useful for research purposes. It is also used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acid and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text