Thermo Scientific Chemicals

4-n-Pentylaniline, 98%, Thermo Scientific Chemicals

Catalog number: H26569.14
25 g, Each
Thermo Scientific Chemicals

4-n-Pentylaniline, 98%, Thermo Scientific Chemicals

Catalog number: H26569.14
25 g, Each
Quantity
Catalog number: H26569.14
also known as H26569-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33228-44-3
IUPAC Name
4-pentylaniline
Molecular Formula
C11H17N
InChI Key
DGFTWBUZRHAHTH-UHFFFAOYSA-N
SMILES
CCCCCC1=CC=C(N)C=C1
Appearance (Color)
Clear colorless to yellow to orange
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.5265-1.5305 @ 20°C
Identification (FTIR)
Conforms

Description

4-Pentylaniline is used in the synthesis of 4-[(4?-pentyl)azo]phenol (5Az-OH) by diazo-coupling reaction with phenol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Pentylaniline is used in the synthesis of 4-[(4′-pentyl)azo]phenol (5Az-OH) by diazo-coupling reaction with phenol

Solubility
Sparingly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text