Thermo Scientific Chemicals

Pyridine-2,6-dicarbonyl dichloride, 97%, Thermo Scientific Chemicals

Catalog number: H26570.14
25 g, Each
Thermo Scientific Chemicals

Pyridine-2,6-dicarbonyl dichloride, 97%, Thermo Scientific Chemicals

Catalog number: H26570.14
25 g, Each
Quantity
Catalog number: H26570.14
also known as H26570-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3739-94-4
IUPAC Name
pyridine-2,6-dicarbonyl dichloride
Molecular Formula
C7H3Cl2NO2
InChI Key
GWHOGODUVLQCEB-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=CC(=N1)C(Cl)=O
Melting Point
57-62?C
Form
Crystals, crystalline powder or fused solid (Chunks and/or Lumps)
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Solution Test
(5% w/v in Acetone) : Clear

Description

2,6-Pyridinedicarbonyl dichloride is used as the reagent during the synthesis of pyridine-based polyamido-polyester optically active macrocycles. It is used as starting reagent during the synthesis of pyridine-bridged 2,6-bis-carboxamide Schiff?s bases. It was used in the synthesis of N,N?-bis(1-pyrenylmethyl)pyridine-2,6-dicarboxamide3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Pyridinedicarbonyl dichloride is used as the reagent during the synthesis of pyridine-based polyamido-polyester optically active macrocycles. It is used as starting reagent during the synthesis of pyridine-bridged 2,6-bis-carboxamide Schiff′s bases. It was used in the synthesis of N,N′-bis(1-pyrenylmethyl)pyridine-2,6-dicarboxamide3

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text