2-(p-Tolyl)pyridine, 98%, Thermo Scientific Chemicals
2-(p-Tolyl)pyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(p-Tolyl)pyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H26574.14
also known as H26574-14
Price (USD)/ Each
183.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
183.00
Add to cart
2-(p-Tolyl)pyridine, 98%, Thermo Scientific Chemicals
Catalog numberH26574.14
Price (USD)/ Each
183.00
-
Add to cart
Chemical Identifiers
CAS4467-06-5
IUPAC Name2-(4-methylphenyl)pyridine
Molecular FormulaC12H11N
InChI KeyKJNZQKYSNAQLEO-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.6150-1.6190 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only