Thermo Scientific Chemicals

Ammonium trifluoroacetate, 98%, Thermo Scientific Chemicals

Catalog number: H26577.22
100 g, Each
Thermo Scientific Chemicals

Ammonium trifluoroacetate, 98%, Thermo Scientific Chemicals

Catalog number: H26577.22
100 g, Each
Quantity
Catalog number: H26577.22
also known as H26577-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3336-58-1
IUPAC Name
ammonium trifluoroacetic acid
Molecular Formula
C2H5F3NO2
InChI Key
YCNIBOIOWCTRCL-UHFFFAOYSA-O
SMILES
[NH4+].OC(=O)C(F)(F)F
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
121-128°C
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

Used in the synthesis of hexafluoro-2-aminopentan-4-one ligand and in tuning and calibrating new liquid chromatography/mass spectrometry systems. Applications of ammonium trifluoroacetate as an additive for elution of chiral acids and bases from derivatized polysaccharide stationary phases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of hexafluoro-2-aminopentan-4-one ligand and in tuning and calibrating new liquid chromatography/mass spectrometry systems. Applications of ammonium trifluoroacetate as an additive for elution of chiral acids and bases from derivatized polysaccharide stationary phases

Solubility
Soluble in Water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture. Keep under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text