Thermo Scientific Chemicals

p-Sexiphenyl, 95%, Thermo Scientific Chemicals

Catalog number: H26586.MD
250 mg, Each
Thermo Scientific Chemicals

p-Sexiphenyl, 95%, Thermo Scientific Chemicals

Catalog number: H26586.MD
250 mg, Each
Quantity
Catalog number: H26586.MD
also known as H26586-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4499-83-6
IUPAC Name
4,4'-bis({[1,1'-biphenyl]-4-yl})-1,1'-biphenyl
Molecular Formula
C36H26
InChI Key
ZEMDSNVUUOCIED-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Assay (unspecified)
≥94.0% (based on elemental C)
Elemental Analysis
H: 5.37-5.82% (Theory: 5.71%)
Appearance (Color)
White to cream or pale yellow
Elemental Analysis
C: 88.63-96.18% (Theory: 94.29%)
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text