Bis(p-toluenesulfonyl)methane, 97%, Thermo Scientific Chemicals
Bis(p-toluenesulfonyl)methane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(p-toluenesulfonyl)methane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H26617.06
also known as H26617-06
Price (USD)/ Each
350.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
350.00
Add to cart
Bis(p-toluenesulfonyl)methane, 97%, Thermo Scientific Chemicals
Catalog numberH26617.06
Price (USD)/ Each
350.00
-
Add to cart
Chemical Identifiers
CAS15310-28-8
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only