2,6-Difluoro-3-methoxyaniline, 97%, Thermo Scientific Chemicals
2,6-Difluoro-3-methoxyaniline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Difluoro-3-methoxyaniline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26631.03
also known as H26631-03
Price (USD)/ Each
213.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
213.00
Add to cart
2,6-Difluoro-3-methoxyaniline, 97%, Thermo Scientific Chemicals
Catalog numberH26631.03
Price (USD)/ Each
213.00
-
Add to cart
Chemical Identifiers
CAS144851-62-7
IUPAC Name2,6-difluoro-3-methoxyaniline
Molecular FormulaC7H7F2NO
InChI KeyXVLHUQRLEPQGDI-UHFFFAOYSA-N
SMILESCOC1=C(F)C(N)=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5210-1.5260 @ 20?C
Appearance (Color)Clear colorless to pale orange
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only