1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane, Thermo Scientific Chemicals
1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
250 mg
Catalog number H26643.MD
also known as H26643-MD
Price (USD)/ Each
190.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
190.00
Add to cart
1,8-Dibenzyl-1,4,8,11-tetraazacyclotetradecane, Thermo Scientific Chemicals
Catalog numberH26643.MD
Price (USD)/ Each
190.00
-
Add to cart
Chemical Identifiers
CAS214078-93-0
IUPAC Name1,8-dibenzyl-1,4,8,11-tetraazacyclotetradecane
Molecular FormulaC24H36N4
InChI KeyQWGFFCRTIFBAFJ-UHFFFAOYSA-N
SMILESC(N1CCCNCCN(CC2=CC=CC=C2)CCCNCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Proton NMRConforms to structure
FormPowder and/or lumps
Carbon-13 NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only