Thermo Scientific Chemicals

(+/-)-Propranolol hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: H26645.14
25 g, Each
Thermo Scientific Chemicals

(+/-)-Propranolol hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: H26645.14
25 g, Each
Quantity
Catalog number: H26645.14
also known as H26645-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
318-98-9
IUPAC Name
hydrogen 1-(naphthalen-1-yloxy)-3-[(propan-2-yl)amino]propan-2-ol chloride
Molecular Formula
C16H22ClNO2
InChI Key
ZMRUPTIKESYGQW-UHFFFAOYNA-N
SMILES
[H+].[Cl-].CC(C)NCC(O)COC1=C2C=CC=CC2=CC=C1
Form
Powder
Assay from Supplier's CofA
≥98.5% (US-sourced material)
Identification (FTIR)
Conforms
Appearance (Color)
White
Comment
Material sourced in UK and US

Description

A beta-adrenergic blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text