2-Acetylquinoxaline is used as dyes, pharmaceuticals, and antibiotics such as olaquindox, carbadox, echinomycin, levomycin and actinoleutin. It is also used to make amiloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Acetylquinoxaline is used as dyes, pharmaceuticals, and antibiotics such as olaquindox, carbadox, echinomycin, levomycin and actinoleutin. It is also used to make amiloride.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
E Abushanab. One-Step Synthesis of Quinoxaline 1,4-Dioxides and Related Compounds. J. Org. Chem. 1970, 35, (12), 4279-4280.
Gerhard Sarodnick.; Matthias Heydenreich.; Torsten Linker.; Erich Kleinpeter. Quinoxalines. Part 12: Synthesis and structural study of 1-(thiazol-2-yl)-1H-pyrazolo[3,4-b]quinoxalines—the dehydrogenative cyclization with hydroxylamine hydrochloride. Tetrahedron. 2003, 59, (33), 6311-6321.