Thermo Scientific Chemicals

4-n-Octylaniline, 99%, Thermo Scientific Chemicals

Catalog number: H26713.06
5 g, Each
Thermo Scientific Chemicals

4-n-Octylaniline, 99%, Thermo Scientific Chemicals

Catalog number: H26713.06
5 g, Each
Quantity
Catalog number: H26713.06
also known as H26713-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16245-79-7
Appearance (Color)
Clear colorless to yellow to orange
Assay (GC)
≥98.5%
Form
Liquid
Refractive Index
1.5145-1.5175 @ 20?C

Description

4-Octylaniline can be been used as an extractant during the extraction of microgram level concentrations of ruthenium(IV) from halide medium. It can also been used in the synthesis of selective benzimidazole based analogs of sphingosine-1-phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Octylaniline can be been used as an extractant during the extraction of microgram level concentrations of ruthenium(IV) from halide medium. It can also been used in the synthesis of selective benzimidazole based analogs of sphingosine-1-phosphate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text