Agar, plant cell culture tested
Agar, plant cell culture tested
Thermo Scientific Chemicals

Agar, plant cell culture tested

Used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number H26724.22
also known as H26724-22
Price (USD)/ Each
88.65
Online exclusive
98.70 
Save 10.05 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
88.65
Online exclusive
98.70 
Save 10.05 (10%)
Add to cart
Agar, plant cell culture tested
Catalog numberH26724.22
Price (USD)/ Each
88.65
Online exclusive
98.70 
Save 10.05 (10%)
-
Add to cart
Chemical Identifiers
CAS9002-18-0
IUPAC Name2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
Molecular FormulaC14H24O9
InChI KeyGYYDPBCUIJTIBM-UHFFFAOYNA-N
SMILESCOC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
CommentMoisture: ≤12%
FormPowder
CommentGel point (1.5%) 34 - 38?C
Particle Size95% is -60 mesh ASTM
View more
Agar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only