Thermo Scientific Chemicals

(S)-(-)-3-Amino-1,2-propanediol, 98%, Thermo Scientific Chemicals

Catalog number: H26743.06
5 g, Each
Thermo Scientific Chemicals

(S)-(-)-3-Amino-1,2-propanediol, 98%, Thermo Scientific Chemicals

Catalog number: H26743.06
5 g, Each
Quantity
Catalog number: H26743.06
also known as H26743-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
61278-21-5
IUPAC Name
3-aminopropane-1,2-diol
Molecular Formula
C3H9NO2
InChI Key
KQIGMPWTAHJUMN-UHFFFAOYNA-N
SMILES
NCC(O)CO
Appearance (Color)
White to yellow
Assay (GC)
≥97.5%
Water Content (Karl Fischer Titration)
≤3.0%
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5% (dry wt. basis)
Optical Rotation
-28? ? 1? (c=4, 5M HCl)

Description

(S)-(-)-3-Amino-1,2-propanediol is used as a chiral selector ligand, used subsequently for a nucleophilic substitution with the dimethoxytrityl (DMT)-protected

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-3-Amino-1,2-propanediol is used as a chiral selector ligand, used subsequently for a nucleophilic substitution with the dimethoxytrityl (DMT)-protected

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text