Thermo Scientific Chemicals

Pyrazine-2-thiocarboxamide, 97%, Thermo Scientific Chemicals

Catalog number: H26750.06
5 g, Each
Thermo Scientific Chemicals

Pyrazine-2-thiocarboxamide, 97%, Thermo Scientific Chemicals

Catalog number: H26750.06
5 g, Each
Quantity
Catalog number: H26750.06
also known as H26750-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4604-72-2
IUPAC Name
pyrazine-2-carbothioamide
Molecular Formula
C5H5N3S
InChI Key
LIURPUMROGYCLW-UHFFFAOYSA-N
SMILES
NC(=S)C1=CN=CC=N1
Appearance (Color)
Yellow
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
195.0-204.0?C
Assay (HPLC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in organic solvents, ethanol, DMSO.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text