General Description
• LB Broth is composed of tryptone (pancreatic digest of casein), sodium chloride, and yeast extract. The tryptone provides nitrogen, vitamins, minerals, and amino acids essential for the growth of these strains. Yeast extract is a source of vitamins, particularly the B-group. The sodium chloride supplies essential electrolytes for transport and osmotic balance
Application
• LB Broth (Lennox) is a rich growth medium that contains all the nutritional requirements for the growth and maintenance of a pure culture of recombinant Escherichia coli strains to allow for plasmid purification and/or protein expression
• LB Broth (Lennox) is used for resuspension of T. pyriformis cell fractions in a study to perform the fractionation of cellular extracts
Literature References
Guennadi Sezonov; Danièle Joseleau-Petit; Richard D'Ari. Escherichia coli Physiology in Luria-Bertani Broth. J. Bacteriole. 2007, 189 (23), 8746-8749.
Neeta L. Lala; Ramakrishnan Ramaseshan; Li Bojun; Subramanian Sundarrajan; R.S. Barhate; Liu Ying-jun; Seeram Ramakrishna. Fabrication of nanofibers with antimicrobial functionality used as filters: protection against bacterial contaminants. Biotechnology and Bioengineering. 2007, 97 (6), 1357-1365.