Thermo Scientific Chemicals

Yeast Extract

Catalog number: H26769.22
100 g, Each
Thermo Scientific Chemicals

Yeast Extract

Catalog number: H26769.22
100 g, Each
Quantity
Catalog number: H26769.22
also known as H26769-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
8013-01-2
Form
Powder
Sodium Chloride
≤0.5%
Total Nitrogen
10-11.8%
Appearance (Color)
Cream to pale yellow to pale brown
Loss on Drying
≤8.0%

Description

Yeast extract is used as food additive, flavor enhancer in processed foods of all kinds and as nutrient for bacterial culture media. It is used to create savoury flavors and umami taste sensations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text