3-(4-Bromophenyl)propionic acid, 98%, Thermo Scientific Chemicals
3-(4-Bromophenyl)propionic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(4-Bromophenyl)propionic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H26785.06
also known as H26785-06
Price (USD)/ Each
102.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
102.00
Add to cart
3-(4-Bromophenyl)propionic acid, 98%, Thermo Scientific Chemicals
Catalog numberH26785.06
Price (USD)/ Each
102.00
-
Add to cart
Chemical Identifiers
CAS1643-30-7
IUPAC Name3-(4-bromophenyl)propanoic acid
Molecular FormulaC9H9BrO2
InChI KeyNCSTWHYWOVZDOC-UHFFFAOYSA-N
SMILESOC(=O)CCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)131.0-137.0?C
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only