6-Methyl-2-pyridinemethanol, 98%, Thermo Scientific Chemicals
6-Methyl-2-pyridinemethanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Methyl-2-pyridinemethanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H26794.14
also known as H26794-14
Price (USD)/ Each
304.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
304.00
Add to cart
6-Methyl-2-pyridinemethanol, 98%, Thermo Scientific Chemicals
Catalog numberH26794.14
Price (USD)/ Each
304.00
-
Add to cart
Chemical Identifiers
CAS1122-71-0
IUPAC Name(6-methylpyridin-2-yl)methanol
Molecular FormulaC7H9NO
InChI KeyJLVBSBMJQUMAMW-UHFFFAOYSA-N
SMILESCC1=CC=CC(CO)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)28.0-34.0?C
FormFused solid
Appearance (Color)White to pale cream
Assay (GC)≥97.5%
Use as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use as primary and secondary intermediate.

Solubility
Low soluble in water, high soluble in ethanol, acetic acid. Soluble in benzene, toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, strong reducing agents. Store under an inert atmosphere.
RUO – Research Use Only

General References:

  1. V. Boekelheide,; W. J. Linn. Rearrangements of N-Oxides. A Novel Synthesis of Pyridyl Carbinols and Aldehydes. J. Am. Chem. Soc. 1954, 76 (5),1286-1291.
  2. P. Vasu Govardhana Reddy,; C. Suresh Reddy,; M. Venugopal. Synthesis and antimicrobial activity of N-substituted N'-[6-methyl-2-oxido-1,3,2-dioxaphosphinino(5,4-b)pyridine-2-yl]ureas. Heteroatom Chemistry. 2003, 14 (6),509-512.