3,5-Bis(trifluoromethyl)thiobenzamide, 97%, Thermo Scientific Chemicals
3,5-Bis(trifluoromethyl)thiobenzamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)thiobenzamide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H26809.MD
also known as H26809-MD
Price (USD)/ Each
60.80
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
60.80
Add to cart
3,5-Bis(trifluoromethyl)thiobenzamide, 97%, Thermo Scientific Chemicals
Catalog numberH26809.MD
Price (USD)/ Each
60.80
-
Add to cart
Chemical Identifiers
CAS317319-15-6
IUPAC Name3,5-bis(trifluoromethyl)benzene-1-carbothioamide
Molecular FormulaC9H5F6NS
InChI KeyOWYYZIVORPIBPZ-UHFFFAOYSA-N
SMILESNC(=S)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)109.0-119.0?C
Appearance (Color)Yellow
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only