Thermo Scientific Chemicals

4-Carboxybenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H26825.06
5 g, Each
Thermo Scientific Chemicals

4-Carboxybenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H26825.06
5 g, Each
Quantity
Catalog number: H26825.06
also known as H26825-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
180516-87-4
IUPAC Name
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Molecular Formula
C13H17BO4
InChI Key
IYDKBQIEOBXLTP-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(O)=O
Melting Point (clear melt)
224.0-234.0?C
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (Silylated GC)
≥96.0%
Form
Crystals or powder or crystalline powder

Description

4-Carboxybenzeneboronic acid pinacol ester is used to improvement of transition metal-mediated carbon-carbon as well as carbon-heteroatom cross-coupling reactions as a labeling feature with the focus on the preparation of radio labeled compounds for molecular imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Carboxybenzeneboronic acid pinacol ester is used to improvement of transition metal-mediated carbon-carbon as well as carbon-heteroatom cross-coupling reactions as a labeling feature with the focus on the preparation of radio labeled compounds for molecular imaging.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text