7-Fluoroisatin, 97%, Thermo Scientific Chemicals
7-Fluoroisatin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Fluoroisatin, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H26827.06
also known as H26827-06
Price (USD)/ Each
77.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
77.00
Add to cart
7-Fluoroisatin, 97%, Thermo Scientific Chemicals
Catalog numberH26827.06
Price (USD)/ Each
77.00
-
Add to cart
Chemical Identifiers
CAS317-20-4
IUPAC Name7-fluoro-2,3-dihydro-1H-indole-2,3-dione
Molecular FormulaC8H4FNO2
InChI KeyHGBGVEOXPHGSOS-UHFFFAOYSA-N
SMILESFC1=CC=CC2=C1NC(=O)C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
Assay (HPLC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)190.0-199.0?C
Identification (FTIR)Conforms
7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.

Solubility
Sparingly Soluble in water (0.022 g/L)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Wendell Wierenga.; John Griffin.; Martha A. Warpehoski. A versatile and efficient process to 3-substituted indoles from anilines. Tetrahedron Letters. 1983, 24, (24), 2437-2440.
  2. Sarah (Sally) L. Price. Computed Crystal Energy Landscapes for Understanding and Predicting Organic Crystal Structures and Polymorphism. Acc. Chem. Res. 2009, 42, (1), 117-126.