Thermo Scientific Chemicals

alpha,alpha-Dimethylbenzylamine, 96%, Thermo Scientific Chemicals

Catalog number: H26845.06
5 g, Each
Thermo Scientific Chemicals

alpha,alpha-Dimethylbenzylamine, 96%, Thermo Scientific Chemicals

Catalog number: H26845.06
5 g, Each
Quantity
Catalog number: H26845.06
also known as H26845-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
585-32-0
Appearance (Color)
Clear colorless
Assay (GC)
≥95.0%
Form
Liquid
Refractive Index
1.5170-1.5230 @ 20?C

Description

Agrochemical Intermediates, dyestuff Intermediate, pharmaceutical Intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agrochemical Intermediates, dyestuff Intermediate, pharmaceutical Intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text