Agarose D1-LE, molecular biology grade
Agarose D1-LE, molecular biology grade
Thermo Scientific Chemicals

Agarose D1-LE, molecular biology grade

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number H26855.36
also known as H26855-36
Price (USD)/ Each
966.65
Online exclusive
1074.00 
Save 107.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
966.65
Online exclusive
1074.00 
Save 107.35 (10%)
Add to cart
Agarose D1-LE, molecular biology grade
Catalog numberH26855.36
Price (USD)/ Each
966.65
Online exclusive
1074.00 
Save 107.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9012-36-6
IUPAC Name(2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC24H38O19
InChI KeyMJQHZNBUODTQTK-WKGBVCLCSA-N
SMILESOC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
CommentGel strength @1% ≥ 1200g/cm²
CommentGel temp 36 ± 1.5°C
Appearance (Color)White
CommentEEO 0.05-0.13
Agarose D1-LE, Molecular Biology Grade is used for molecular biology applications. It acts as the medium for analytical scale electrophoretic separation in agarose gel electrophoresis. It is suitable for analytical and preparative nucleic acid electrophoresis, blotting and protein electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose D1-LE, Molecular Biology Grade is used for molecular biology applications. It acts as the medium for analytical scale electrophoretic separation in agarose gel electrophoresis. It is suitable for analytical and preparative nucleic acid electrophoresis, blotting and protein electrophoresis.

Solubility
Soluble in water.

Notes
Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. A general-purpose agarose for molecular biology applications. It has a low electroendosmosis(EEO) and high electrophoresis mobility. Suitable for analytical and preparative nucleic acid electrophoresis, blotting and protein electrophoresis.
  2. Gómez-Mascaraque, L. G.; Méndez, J. A.; Fernández-Gutiérrez, M.; Vázquez, B.; San Román, J. Oxidized dextrins as alternative crosslinking agents for polysaccharides: Application to hydrogels of agarose-chitosan. Acta Biomater. 2014, 10 (2), 798-811.
  3. Natoli, A.; Cabeza, A.; Torre, Á. G.; Aranda, M. A.; Santacruz, I. Colloidal processing of macroporous TiO2 Materials for photocatalytic water treatment. J. Am. Ceram. Soc. 2012, 95 (2), 502-508.
  4. Geng, T.; Mathies, R. A. Minimizing inhibition of PCR-STR typing using digital agarose droplet microfluidics. Forensic Sci. Int. Genet. 2015, 14, 203-209.