Thermo Scientific Chemicals

Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 94%, Thermo Scientific Chemicals

Catalog number: H26857.14
25 g, Each
Thermo Scientific Chemicals

Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 94%, Thermo Scientific Chemicals

Catalog number: H26857.14
25 g, Each
Quantity
Catalog number: H26857.14
also known as H26857-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
120801-75-4
IUPAC Name
trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate
Molecular Formula
C5H9F3O4SSi
InChI Key
XHVSCKNABCCCAC-UHFFFAOYSA-N
SMILES
C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O
Appearance (Color)
Clear, colorless to pale yellow
Fluorine-19 NMR
≥92.5%
Refractive Index
1.3635-1.3705 @ 20?C
Form
Liquid

Description

Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate is an fluoroalkylation agent, reactant for preparation of difluorocyclopropenes, preparation of difluorocarbene reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate is an fluoroalkylation agent, reactant for preparation of difluorocyclopropenes, preparation of difluorocarbene reagents.

Solubility
Reacts with water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text