Vinyloxytrimethylsilane, 97%, Thermo Scientific Chemicals
Vinyloxytrimethylsilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vinyloxytrimethylsilane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26876.03
also known as H26876-03
Price (USD)/ Each
33.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
33.20
Add to cart
Vinyloxytrimethylsilane, 97%, Thermo Scientific Chemicals
Catalog numberH26876.03
Price (USD)/ Each
33.20
-
Add to cart
Chemical Identifiers
CAS6213-94-1
IUPAC Name(ethenyloxy)trimethylsilane
Molecular FormulaC5H12OSi
InChI KeyHFTNNOZFRQLFQB-UHFFFAOYSA-N
SMILESC[Si](C)(C)OC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Solution TestClear (10% in methanol)
FormLiquid
Solution TestClear (10% in diethyl ether)
Assay (GC)≥96.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only