Thermo Scientific Chemicals

Vinyloxytrimethylsilane, 97%, Thermo Scientific Chemicals

Catalog number: H26876.06
5 g, Each
Thermo Scientific Chemicals

Vinyloxytrimethylsilane, 97%, Thermo Scientific Chemicals

Catalog number: H26876.06
5 g, Each
Quantity
Catalog number: H26876.06
also known as H26876-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6213-94-1
IUPAC Name
(ethenyloxy)trimethylsilane
Molecular Formula
C5H12OSi
InChI Key
HFTNNOZFRQLFQB-UHFFFAOYSA-N
SMILES
C[Si](C)(C)OC=C
Appearance (Color)
Clear colorless to pale yellow
Solution Test
Clear (10% in methanol)
Form
Liquid
Solution Test
Clear (10% in diethyl ether)
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text