Thermo Scientific Chemicals

1-Methylindazole-3-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H26879.03
1 g, Each
Thermo Scientific Chemicals

1-Methylindazole-3-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H26879.03
1 g, Each
Quantity
Catalog number: H26879.03
also known as H26879-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50890-83-0
IUPAC Name
1-methyl-1H-indazole-3-carboxylic acid
Molecular Formula
C9H8N2O2
InChI Key
OVVDFORZEGKEJM-UHFFFAOYSA-N
SMILES
CN1N=C(C(O)=O)C2=CC=CC=C12
Form
Powder
Assay (Silylated GC)
≥96.0%
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

1-Methylindazole-3-carboxylic acid is used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a pharmaceutical adjuvant, Granisetron Impurity D.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylindazole-3-carboxylic acid is used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a pharmaceutical adjuvant, Granisetron Impurity D.

Solubility
Insoluble in water. Soluble in DMSO and Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text