Thermo Scientific Chemicals

Methylamine, 2M in methanol, Thermo Scientific Chemicals

Catalog number: H26889.AE
100 mL, Each
Thermo Scientific Chemicals

Methylamine, 2M in methanol, Thermo Scientific Chemicals

Catalog number: H26889.AE
100 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: H26889.AE
also known as H26889-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
74-89-5
IUPAC Name
methanamine
Molecular Formula
CH5N
InChI Key
BAVYZALUXZFZLV-UHFFFAOYSA-N
SMILES
CN
Form
liquid
Appearance (Color)
Clear, colourless
Assay (Aqueous acid-base Titration)
1.90-2.10M
Refractive Index
1.3340-1.3370

Description

Used for making pharmaceuticals, paint removers, surfactants, rubber chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text