3,5-Dimethylisoxazole-4-boronic acid, 97%, Thermo Scientific Chemicals
3,5-Dimethylisoxazole-4-boronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dimethylisoxazole-4-boronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26893.03
also known as H26893-03
Price (USD)/ Each
109.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
109.00
Add to cart
3,5-Dimethylisoxazole-4-boronic acid, 97%, Thermo Scientific Chemicals
Catalog numberH26893.03
Price (USD)/ Each
109.00
-
Add to cart
Chemical Identifiers
CAS16114-47-9
IUPAC Name(3,5-dimethyl-1,2-oxazol-4-yl)boronic acid
Molecular FormulaC5H8BNO3
InChI KeyDIIFZCPZIRQDIJ-UHFFFAOYSA-N
SMILESCC1=C(B(O)O)C(C)=NO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Assay (Aqueous acid-base Titration)≥96.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only