Diethyl acetylmethylphosphonate, 97%, Thermo Scientific Chemicals
Diethyl acetylmethylphosphonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl acetylmethylphosphonate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H26896.06
also known as H26896-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1067-71-6
IUPAC Namediethyl (2-oxopropyl)phosphonate
Molecular FormulaC7H15O4P
InChI KeyRSAFKRSMGOSHRK-UHFFFAOYSA-N
SMILESCCOP(=O)(CC(C)=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4315-1.4365 @ 20?C
Diethyl acetylmethylphosphonate is used as ligand in coordination chemistry to prepare lanthanide series metals like terbium, europium and neodymium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl acetylmethylphosphonate is used as ligand in coordination chemistry to prepare lanthanide series metals like terbium, europium and neodymium.

Solubility
Miscible with tetrahydrofuran, ether, dichloromethane and chloroform.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Iranpoor, N.; Firouzabadi, H.; Moghadam, K. R.; Davan, E. E. Triphenylphosphine/2, 3-Dichloro-5, 6-dicyanobenzoquinone (PPh3/DDQ) System for Conversion of Alcohols and Thiols into Trialkyl Phosphonates. Asian J. Org. Chem. 2015, 4 (11), 1289-1293.
  2. Xiong, F.; Wang, H.; Yan, L.; Xu, L.; Tao, Y.; Wu, Y.; Chen, F. Diastereoselective synthesis of pitavastatin calcium via bismuth-catalyzed two-component hemiacetal/oxa-Michael addition reaction. Org. Biomol. Chem. 2015, 13 (38), 9813-9819.