Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium dicyanamide, 98%, Thermo Scientific Chemicals

Catalog number: H26901.06
5 g, Each
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium dicyanamide, 98%, Thermo Scientific Chemicals

Catalog number: H26901.06
5 g, Each
Quantity
Catalog number: H26901.06
also known as H26901-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
370865-89-7
IUPAC Name
1-ethyl-3-methyl-1H-imidazol-3-ium; [(cyanoimino)methylidene]azanide
Molecular Formula
C8H11N5
InChI Key
MKHFCTXNDRMIDR-UHFFFAOYSA-N
SMILES
[N-]=C=NC#N.CCN1C=C[N+](C)=C1
Appearance (Color)
Clear colorless to yellow to orange-brown
Assay from Suppliers CofA
≥97.5%
Refractive Index
1.5130-1.5170 @ 20°C
Form
Liquid

Description

Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.

Solubility
Soluble in water. Soluble in solvents like methanol, acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text