Chemical Identifiers
CAS85959-83-7
IUPAC Namehafnium(4+) bis(pentamethylcyclopenta-1,3-dien-1-ide) dichloride
Molecular FormulaC20H30Cl2Hf
InChI KeyYPRLSLCBPOTFGE-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Hf+4].CC1=[C-]C(C)(C)C(C)=C1C.CC1=[C-]C(C)(C)C(C)=C1C
View more