N(alpha)-Boc-L-2,3-diaminopropionic acid, 97%, Thermo Scientific Chemicals
N(alpha)-Boc-L-2,3-diaminopropionic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N(alpha)-Boc-L-2,3-diaminopropionic acid, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Catalog number H26919.03
also known as H26919-03
Price (USD)/ Each
73.90
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
73.90
Add to cart
N(alpha)-Boc-L-2,3-diaminopropionic acid, 97%, Thermo Scientific Chemicals
Catalog numberH26919.03
Price (USD)/ Each
73.90
-
Add to cart
Chemical Identifiers
CAS73259-81-1
IUPAC Name(2S)-3-azaniumyl-2-{[(tert-butoxy)carbonyl]amino}propanoate
Molecular FormulaC8H16N2O4
InChI KeyKRJLRVZLNABMAT-YFKPBYRVSA-N
SMILESCC(C)(C)OC(=O)N[C@@H](C[NH3+])C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Optical Rotation-13.5 ± 1.5? (C=1 in water)
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only