Thermo Scientific Chemicals

3-Bromocatechol, 95%, Thermo Scientific Chemicals

Catalog number: H26925.MD
250 mg, Each
Thermo Scientific Chemicals

3-Bromocatechol, 95%, Thermo Scientific Chemicals

Catalog number: H26925.MD
250 mg, Each
Quantity
Catalog number: H26925.MD
also known as H26925-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14381-51-2
IUPAC Name
3-bromobenzene-1,2-diol
Molecular Formula
C6H5BrO2
InChI Key
JPBDMIWPTFDFEU-UHFFFAOYSA-N
SMILES
OC1=CC=CC(Br)=C1O
Appearance (Color)
White to cream or pale yellow
Assay (GC)
≥94.0%
Melting Point (clear melt)
36-45?C
Water Content (Karl Fischer Titration)
<2.0%
Form
Crystalline or lumpy powder or crystals

Description

3-Bromocatechol, is used as a drug and agrochemicals intermediate, Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromocatechol, is used as a drug and agrochemicals intermediate, Pharmaceutical intermediate.

Solubility
Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text