4-Bromo-o-phenylenediamine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromo-o-phenylenediamine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only
MA Phillips. The methylation of benziminazoles. Journal of the Chemical Society. 193111 (1), 1143-1153.
EL Samuel. Nitration and bromination products of 2-trifluoromethylbenzimidazoles and their NMR shift assignments. Australian Journal of Chemistry.197225 (12), 2725 - 2729 .