Thermo Scientific Chemicals

4-Bromo-o-phenylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: H26929.22
100 g, Each
Thermo Scientific Chemicals

4-Bromo-o-phenylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: H26929.22
100 g, Each
Quantity
Catalog number: H26929.22
also known as H26929-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1575-37-7
Appearance (Color)
Pale brown to brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
59.0-68.0?C

Description

4-Bromo-o-phenylenediamine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-o-phenylenediamine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text