Thermo Scientific Chemicals

(R)-(-)-3-Amino-1-Boc-piperidine, 98%, Thermo Scientific Chemicals

Catalog number: H26937.06
5 g, Each
Thermo Scientific Chemicals

(R)-(-)-3-Amino-1-Boc-piperidine, 98%, Thermo Scientific Chemicals

Catalog number: H26937.06
5 g, Each
Quantity
Catalog number: H26937.06
also known as H26937-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
188111-79-7
IUPAC Name
tert-butyl 3-aminopiperidine-1-carboxylate
Molecular Formula
C10H20N2O2
InChI Key
AKQXKEBCONUWCL-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)N1CCCC(N)C1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4710-1.4750 @ 20?C
Optical Rotation
-27.5 ± 1.5? (neat)

Description

(R)-(-)-3-Amino-1-Boc-piperidine is used as an intermediate in the manufacture of various substances such as pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-3-Amino-1-Boc-piperidine is used as an intermediate in the manufacture of various substances such as pharmaceuticals.

Solubility
Immiscible with water.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text