(+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, 99%, Thermo Scientific Chemicals
(+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H26949.03
also known as H26949-03
Price (USD)/ Each
34.70
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
34.70
Add to cart
(+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, 99%, Thermo Scientific Chemicals
Catalog numberH26949.03
Price (USD)/ Each
34.70
-
Add to cart
Chemical Identifiers
CAS101203-31-0
IUPAC Name3,3'-di-tert-butyl-5,5',6,6'-tetramethyl-[1,1'-biphenyl]-2,2'-diol
Molecular FormulaC24H34O2
InChI KeyNMVVBVMYPLMIOU-UHFFFAOYSA-N
SMILESCC1=CC(=C(O)C(=C1C)C1=C(C)C(C)=CC(=C1O)C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)161.5-167.5?C
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only