Chlorobis(4-fluorophenyl)phosphine, 98%, Thermo Scientific Chemicals
Chlorobis(4-fluorophenyl)phosphine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chlorobis(4-fluorophenyl)phosphine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number H26953.06
also known as H26953-06
Price (USD)/ Each
745.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
745.00
Add to cart
Chlorobis(4-fluorophenyl)phosphine, 98%, Thermo Scientific Chemicals
Catalog numberH26953.06
Price (USD)/ Each
745.00
-
Add to cart
Chemical Identifiers
CAS23039-97-6
IUPAC Namechlorobis(4-fluorophenyl)phosphane
Molecular FormulaC12H8ClF2P
InChI KeyPYFWYPBHSZATST-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)P(Cl)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Phosphorus-31 NMRConforms
Assay (GC)≥97.5%
In chemical research. Used as an interphasic transfer catalyst, a polymerization catalyst, and a trans-halogenation catalyst. As a precursor to fire-retardant materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In chemical research. Used as an interphasic transfer catalyst, a polymerization catalyst, and a trans-halogenation catalyst. As a precursor to fire-retardant materials.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, moisture.
RUO – Research Use Only

General References:

  1. Laura A. Rusch-Salazar,; Valerie V. Sheare. A series of new high-performance materials based on poly[4'-fluorophenyl-bis(4-phenyl)phosphine oxide]. Journal of Polymer Science Part A: Polymer Chemistry. 2003, 41 (14),2277-2287.