Thermo Scientific Chemicals

Chlorobis(4-fluorophenyl)phosphine, 98%, Thermo Scientific Chemicals

Catalog number: H26953.MD
250 mg, Each
Thermo Scientific Chemicals

Chlorobis(4-fluorophenyl)phosphine, 98%, Thermo Scientific Chemicals

Catalog number: H26953.MD
250 mg, Each
Quantity
Catalog number: H26953.MD
also known as H26953-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23039-97-6
IUPAC Name
chlorobis(4-fluorophenyl)phosphane
Molecular Formula
C12H8ClF2P
InChI Key
PYFWYPBHSZATST-UHFFFAOYSA-N
SMILES
FC1=CC=C(C=C1)P(Cl)C1=CC=C(F)C=C1
Appearance (Color)
Clear colorless
Form
Liquid
Phosphorus-31 NMR
Conforms
Assay (GC)
≥97.5%

Description

In chemical research. Used as an interphasic transfer catalyst, a polymerization catalyst, and a trans-halogenation catalyst. As a precursor to fire-retardant materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In chemical research. Used as an interphasic transfer catalyst, a polymerization catalyst, and a trans-halogenation catalyst. As a precursor to fire-retardant materials.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text