Thermo Scientific Chemicals

2-Cyano-3-iodopyridine, 95%, Thermo Scientific Chemicals

Catalog number: H26963.MD
250 mg, Each
Thermo Scientific Chemicals

2-Cyano-3-iodopyridine, 95%, Thermo Scientific Chemicals

Catalog number: H26963.MD
250 mg, Each
Quantity
Catalog number: H26963.MD
also known as H26963-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
827616-52-4
IUPAC Name
3-iodopyridine-2-carbonitrile
Molecular Formula
C6H3IN2
InChI Key
MZZGCCSFZCRVAF-UHFFFAOYSA-N
SMILES
IC1=C(N=CC=C1)C#N
Assay (GC)
≥94.0%
Appearance (Color)
White to pale cream
Melting Point (clear melt)
94.5-103.5?C
Form
Powder

Description

Used to prepare Grignard reagents which are widely used in organic synthesis.2-Cyano-3-iodopyridine are used in coupling reactions. Has wide application in the pharmaceutical as well as chemical industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to prepare Grignard reagents which are widely used in organic synthesis.2-Cyano-3-iodopyridine are used in coupling reactions. Has wide application in the pharmaceutical as well as chemical industries.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text