Thermo Scientific Chemicals

2-Chloro-6-methoxyquinoline-3-carboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: H26973.06
5 g, Each
Thermo Scientific Chemicals

2-Chloro-6-methoxyquinoline-3-carboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: H26973.06
5 g, Each
Quantity
Catalog number: H26973.06
also known as H26973-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
73568-29-3
IUPAC Name
2-chloro-6-methoxyquinoline-3-carbaldehyde
Molecular Formula
C11H8ClNO2
InChI Key
TZQOMBXDCIPJKW-UHFFFAOYSA-N
SMILES
COC1=CC=C2N=C(Cl)C(C=O)=CC2=C1
Appearance (Color)
Yellow to pale orange or pale brown
Assay (GC)
≥98.5%
Free acid (titration)
≤0.5%
Melting Point (clear melt)
145-151?C
Form
Powder

Description

Mainly used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings. As raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mainly used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings. As raw material in organic synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text