Mainly used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings. As raw material in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Mainly used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings. As raw material in organic synthesis.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only
Subashini,; S. Mohana Roopan,; F. nawaz khan. Synthesis and Free Radical Scavenging Property of some Quinoline Derivatives. J. Chil. Chem. Soc. 2010, 55 (3),317-319.
F. N. Khan,; R. Subashini,; S. M. Roopan,; V. R. Hathwar,;S. W. Ng. 2-Chloro-6-methylquinoline-3-carbaldehyde . Journal name. year of publication , volume issue , pg no.