Thermo Scientific Chemicals

1-n-Butyl-3-methylpyridinium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog number: H26983.18
50 g, Each
Thermo Scientific Chemicals

1-n-Butyl-3-methylpyridinium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog number: H26983.18
50 g, Each
Quantity
Catalog number: H26983.18
also known as H26983-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
597581-48-1
IUPAC Name
1-butyl-3-methylpyridin-1-ium; tetrafluoroboranuide
Molecular Formula
C10H16BF4N
InChI Key
UQEAMJVKASNOMA-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.CCCC[N+]1=CC=CC(C)=C1
Appearance (Color)
Clear, colourless to brown
Form
liquid
Assay (unspecified)
> 98.5%
Water Content (Karl Fischer Titration)
< 1.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text