Thermo Scientific Chemicals

(+/-)-2,2'-Dibromo-1,1'-binaphthyl, 96%, Thermo Scientific Chemicals

Catalog number: H26987.06
5 g, Each
Thermo Scientific Chemicals

(+/-)-2,2'-Dibromo-1,1'-binaphthyl, 96%, Thermo Scientific Chemicals

Catalog number: H26987.06
5 g, Each
Quantity
Catalog number: H26987.06
also known as H26987-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
74866-28-7
IUPAC Name
2,2'-dibromo-1,1'-binaphthalene
Molecular Formula
C20H12Br2
InChI Key
IJUDEFZBMMRSNM-UHFFFAOYSA-N
SMILES
BrC1=CC=C2C=CC=CC2=C1C1=C(Br)C=CC2=CC=CC=C12
Form
Powder
Identification (FTIR)
Conforms
Appearance (Color)
White to cream
Assay (GC)
≥95.0%
Melting Point (clear melt)
177-183?C

Description

The synthesis of this class of compounds relies on the lithiation of racemic 2,2'-dibromo-1,1'-binaphthyl with t-BuLi followed by addition of Ph 2 PCl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The synthesis of this class of compounds relies on the lithiation of racemic 2,2′-dibromo-1,1′-binaphthyl with t-BuLi followed by addition of Ph 2 PCl.

Solubility
Sparingly Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text