1-n-Butylpyridinium bromide, 99%, Thermo Scientific Chemicals
1-n-Butylpyridinium bromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-n-Butylpyridinium bromide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number H26995.06
also known as H26995-06
Price (USD)/ Each
50.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
50.30
Add to cart
1-n-Butylpyridinium bromide, 99%, Thermo Scientific Chemicals
Catalog numberH26995.06
Price (USD)/ Each
50.30
-
Add to cart
Chemical Identifiers
CAS874-80-6
IUPAC Name1-butylpyridin-1-ium bromide
Molecular FormulaC9H14BrN
InChI KeyKVBQNFMTEUEOCD-UHFFFAOYSA-M
SMILES[Br-].CCCC[N+]1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)103.0-109.0?C
Assay (Titration ex Bromide)≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)≤0.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder or lumps
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only