Thermo Scientific Chemicals

1-n-Butylpyridinium bromide, 99%, Thermo Scientific Chemicals

Catalog number: H26995.18
50 g, Each
Thermo Scientific Chemicals

1-n-Butylpyridinium bromide, 99%, Thermo Scientific Chemicals

Catalog number: H26995.18
50 g, Each
Quantity
Catalog number: H26995.18
also known as H26995-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
874-80-6
IUPAC Name
1-butylpyridin-1-ium bromide
Molecular Formula
C9H14BrN
InChI Key
KVBQNFMTEUEOCD-UHFFFAOYSA-M
SMILES
[Br-].CCCC[N+]1=CC=CC=C1
Melting Point (clear melt)
103.0-109.0?C
Assay (Titration ex Bromide)
≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)
≤0.5%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or lumps

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text