Thermo Scientific Chemicals

6-Methylchromone, 98%, Thermo Scientific Chemicals

Catalog number: H27002.06
5 g, Each
Thermo Scientific Chemicals

6-Methylchromone, 98%, Thermo Scientific Chemicals

Catalog number: H27002.06
5 g, Each
Quantity
Catalog number: H27002.06
also known as H27002-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38445-23-7
IUPAC Name
6-methyl-4H-chromen-4-one
Molecular Formula
C10H8O2
InChI Key
HTXQVFXXVXOLCF-UHFFFAOYSA-N
SMILES
CC1=CC=C2OC=CC(=O)C2=C1
Appearance (Color)
Yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
86.5-92.5?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text