Thermo Scientific Chemicals

1-Boc-3-pyrrolidinone, 97%, Thermo Scientific Chemicals

Catalog number: H27023.03
1 g, Each
Thermo Scientific Chemicals

1-Boc-3-pyrrolidinone, 97%, Thermo Scientific Chemicals

Catalog number: H27023.03
1 g, Each
Quantity
Catalog number: H27023.03
also known as H27023-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
101385-93-7
IUPAC Name
tert-butyl 3-oxopyrrolidine-1-carboxylate
Molecular Formula
C9H15NO3
InChI Key
JSOMVCDXPUXKIC-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N1CCC(=O)C1
Appearance (Color)
White to cream to dark brown to pale cream
Form
Powder or fused solid
Assay (GC)
≥96.0%
Melting Point (clear melt)
31.0-41.0?C

Description

In the Synthesis and antibacterial evaluation of a novel series of rifabutin-like spirorifamycins 1-Boc-3-pyrrolidinone is used.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In the Synthesis and antibacterial evaluation of a novel series of rifabutin-like spirorifamycins 1-Boc-3-pyrrolidinone is used.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text